missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate (Na2EDTA·2H2O)

Product Code. 11588746
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11588746 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11588746 Supplier Invitrogen™ Supplier No. 15576028

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Suitable for biochemistry or molecular biology applications

  • UltraPure Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate (Na2EDTA•2H2O), is a chelator of divalent metal cation

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material EDTA
Content And Storage Store at room temperature.
For Use With (Application) Biochemistry, Molecular Biology
Product Line UltraPure
Product Type Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate
Quantity 500 g
Purity Molecular Biology Grade
Shipping Condition Room Temperature
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.