missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Acrylamide

Product Code. 11598736
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11598736 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11598736 Supplier Invitrogen™ Supplier No. 15512023
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Recrystallized three times to ensure highly purified product

UltraPure™ Acrylamide is major ingredient in polyacrylamide gels, which are used for electrophoretic separation of both nucleic acid fragments and proteins. DNA fragments generated by restriction endonuclease digestion, dideoxy sequencing, or Maxam and Gilbert sequencing methods can be separated by polyacrylamide gel electrophoresis (PAGE).

  • Proteins can be separated by variety of techniques with polyacrylamide gels, including denaturing gels using SDS or urea, isoelectric focusing gels, and native gels in a wide variety of buffers
  • Performance and Quality Testing: Polymerization time is evaluated in an SDS polyacrylamide gel system

Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Gel Casting, Nucleic Acid Gel Electrophoresis & Blotting, Protein Gel Electrophoresis, Proteins, Expression, Isolation & Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) Gel Electrophoresis
Quantity 500 g
Shipping Condition Room Temperature
Product Line UltraPure
Product Type Acrylamide
compliance-icons compliance-icons
Product Identifier
  • UltraPure Acrylamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H372-Causes damage to organs.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Acrylamide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.