Learn More
Invitrogen™ TrackIt™ 10 bp DNA Ladder
Pre-mixed with sample buffer for ready-to-load convenience
Brand: Invitrogen™ 10488019
Code : 50
Additional Details : Weight : 0.02000kg
Includes: TrackIt™ 10bp DNA Ladder is supplied at 0.5μg/μL in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.4% Orange G, and 5% glycerol; For 20 applications
Description
TrackIt™ 10bp DNA Ladder consists of thirty-three 10bp repeats and is suitable for sizing DNA from 10bp to 330bp. The 100bp band is two- to three- times brighter than the other bands in the ladder, and can be used for orientation. This ladder can be stained with SYBR™ Safe, ethidium bromide (and other nucleic acid stains) or radiolabeled using T4 polynucleotide kinase.
- TrackIt sample buffers are formulated for easy tracking of DNA migration with minimal masking of DNA bands
- Compatible with agarose and E-Gel™ gels
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice
Specifications
0.5 μg/μL | |
Liquid | |
20 Applications |
DNA Molecular Weight Marker/Ladder | |
TrackIt™ 10bp DNA Ladder is supplied at 0.5μg/μL in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.4% Orange G, and 5% glycerol | |
10 to 200 bp |
For Research Use Only. Not for use in diagnostic procedures.