missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOPO™ TA Cloning™ Kit for Subcloning, with One Shot™ TOP10 Electrocomp™ E. coli

Product Code. 10125082
Change view
Click to view available options
Quantity:
25 Reactions
50 Reactions
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10125082 25 Reactions Each
10644223 50 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10125082 Supplier Invitrogen™ Supplier No. K456001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Topoisomerase I-activated pCR™2.1-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Control PCR primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid

Provides a highly efficient, 5-minute, one-step cloning strategy (TOPO Cloning) for the direct insertion of Taq polymerase–amplified PCR products into a plasmid vector for subcloning

Each kit uses the pCR 2.1-TOPO TA vector with convenient restriction sites for subcloning. The cloning kits are available with a variety of competent cells, or without competent cells, depending upon your needs and budget. Each kit uses the pCR™ 2.1-TOPO TA vector with convenient restriction sites for subcloning.
pCR 2.1-TOPO TA vector: cloning and subcloning simplicity
The pCR 2.1-TOPO TA vector is linearized with 3'-thymidine (T) overhangs for direct ligation of Taq polymerase–amplified PCR products (TA cloning) and is “activated” with covalently bound Topoisomerase I. There is no need to add ligase and cloning is complete in 5 minutes. EcoRI sites flanking the PCR product insertion site allow for easy excision of inserts or use any combination of 15 convenient restriction sites flanking your PCR insert for easy, directional subcloning.
pCR 2.1-TOPO TA clone selection and manipulation
The pCR 2.1-TOPO TA vector contains both ampicillin and kanamycin resistance markers and the LacZα gene for blue/white screening.
Simplified TOPO -based cloning
Using TOPO cloning technology, there is no need for PCR primers containing specific sequences, post-PCR procedures, vector preparation, or other time-intensive DNA manipulation steps. Just add your PCR reaction straight to the provided topoisomerase-charged vector, incubate 5 minutes, and transform E. coli competent cells.
Efficient cloning
With up to 95% of clones carrying the desired insert, you can screen less clones, saving time and money. The pCR 2.1-TOPO TA vector used in this kit comes with 3'-T overhangs for efficient ligation of Taq polymerase–amplified PCR products, which contain 3'-A overhangs.
The standard in cloning
When it comes to cloning, TOPO cloning technology has been a reliable partner for thousands of scientists for over ten years. Fast, simple-to-use, and efficient, TOPO cloning has been applied to many different vectors for a wide array of applications.
TOPO TA Cloning Kits for Subcloning: kit options
The TOPO TA Cloning Kit for Sequencing can be purchased with a variety of competent cells that deliver different advantages depending upon your needs:

  • General cloning: TOP10 cells (Cat. No. K4500-01, K4500-40)
  • High-efficiency cloning: TOP10 Electrocomp™ Cells (Cat. No. K4560-01, K4560-40)
  • General cloning, bacteriophage T1 resistance: DH5α-T1R cells (Cat. No. K4520-01, K4520-40)
  • Fast growth: Mach1™ -T1R Chemically Competent E. Coli (Cat. No. K4510-20)
  • Provide your own: for flexibility and to save money (Cat. No. 450641)
Two versions of the kit that include a PureLink™ Quick Plasmid Miniprep Kit (Cat. No. K4500-02 and K4510-02) for use in isolation of clean, sequencing-ready, recombinant plasmid are also available.

  • Fast and easy: go from PCR to clones in just three steps and in as little as 5 minutes hands-on time
  • Efficient: obtain up to 95% clones with correct insert
  • Proven: reliable performance for over a decade with over 4,000 citations
  • Simple: no ligase, post-PCR procedures, or PCR primers containing specific sequences are required
  • Vector: pCR 2.1-TOPO TA vector: subcloning vector with 15 convenient restriction sites flanking your insert for easy, directional subcloning
  • Cloning method: TOPO TA Cloning : Topoisomerase I–based, 5-minute ligation of PCR products with 3′-A overhangs (Taq polymerase amplified) to TOPO vector with T overhangs
  • Competent cells: various options: choose from kits with either general, high-efficiency, bacteriophage T1–resistant, fast-growing competent cells, or use your own

Cloning, PCR Cloning

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain TOP10
Cloning Method TOPO™-TA
Content And Storage Box 1:
• Topoisomerase I-activated pCR™2.1-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid

Store at -68 to -85°C.
Format Kit
Product Line One Shot
Product Type Cloning Kit
Quantity 25 Reactions
Vector TOPO-TA Cloning Vectors

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.