missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOPO™ TA Cloning™ Kit for Sequencing, with One Shot™ TOP10 Electrocomp™ E. coli

Product Code. p-4921691
Click to view available options
Quantity:
25 Reactions
50 Reactions
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10134462

Brand: Invitrogen™ K458001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Topoisomerase I-activated pCR™4-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; T3, T7, M13 forward, and M13 reverse primers; Control PCR primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid

Provides a highly efficient, 5 minute, one-step cloning strategy (TOPO Cloning) for the direct insertion of Taq polymerase-amplified PCR products into a plasmid vector for sequencing

Each kit uses the pCR™4-TOPO™ TA vector with specially designed sequencing primer sites that return more insert sequence and less vector sequence from each reaction. Kit includes everything necessary to clone and select recombinant vectors containing your PCR fragment of choice.
pCR4-TOPO TA Vector: Optimized for Sequencing
We have removed much of the multiple cloning site from the pCR4-TOPO TA vector to shorten the distance between sequencing primer sites and the insert site to as little as 33 bp. This means sequencing reactions give less vector sequence and more insert sequence. The pCR4-TOPO TA vector has sites for four common sequencing primers: M13 forward, M13 reverse, T7, and T3. The kits include an aliquot of each.
pCR4-TOPO TA Clone Selection and Manipulation
The pCR4-TOPO TA vector contains both ampicillin and kanamycin resistance markers and a LacZα-ccdB gene fusion for positive selection and blue/white screening. The vector's minimized multiple cloning site still includes flanking EcoRI sites for simplified excision of cloned PCR products and a unique Sse8387I site for generation of nested deletions prior to sequencing. T7 and T3 promoters are also present for in vitro transcription.
Simplified TOPO-Based Cloning
Using TOPO cloning technology, there is no need for PCR primers containing specific sequences, post-PCR procedures, vector preparation, or other time-intensive DNA manipulation steps. Just add your PCR reaction straight to the provided topoisomerase-charged vector, incubate 5 minutes, and transform with the providedE. coli competent cells.
Efficient Cloning
With up to 95% of clones carrying the desired insert, you can screen less clones to help you save time and money. The pCR4-TOPO TA vector used in this kit comes with 3'T overhangs for efficient ligation of Taq-amplified PCR products with 3'A overhangs.
The Standard in Cloning
When it comes to cloning, TOPO™ cloning technology has been a reliable partner for thousands of scientists for over ten years. Fast, simple-to-use, and efficient, TOPO™ cloning has been applied to many different vectors for a wide array of applications.
TOPO TA Cloning Kits for Sequencing Kit Options
The TOPO TA Cloning Kit for Sequencing can be purchased with a variety of competent cells that deliver different advantages depending on what your needs are:

  • General cloning: TOP10 (Cat. No. K4575-J10, K4575-01, K4575-40)
  • High efficiency cloning: TOP10 Electrocomp™ Cells (Cat. No. K4580-01, K4580-40)
  • General cloning, bacteriophage T1 resistance: DH5α-T1R (Cat. No. K4595-01, K4595-40)
  • Fast growth: Mach1™-T1R Chemically Competent E. Coli (Cat. No. K4530-20)
A version of the kit that includes a PureLink™ Quick Plasmid Miniprep Kit (Cat. No. K4575-02) for use in isolation of clean, sequencing-ready, recombinant plasmid is also available.

  • Get More Sequence: Allows for more insert sequence and less vector sequence when using standard sequencing primers
  • Fast and Easy: Go from PCR-to-clones in just three steps and in as little as 5 minutes hands-on time
  • Efficient: Achieve up to 95% clones with correct insert
  • Proven: Reliable performance for over a decade with over 4000 citations
  • VECTOR: pCR4-TOPO TA Vector: Optimized cloning vector for improved sequencing results
  • CLONING METHOD: TOPO TA Cloning: Topoisomerase I based 5 minute ligation of PCR products with 3'A overhangs (Taq-amplified) to the vector
  • COMPETENT CELLS: Various Options: Choose from kits with either general, high-efficiency, bacteriophage T1-resistant, or fast-growing competent cells

Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Cloning, Methylation Analysis, PCR Cloning, RNAi, Epigenetics and Non-Coding RNA Research, Sequencing Bisulfite-Treated DNA

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain TOP10
Cloning Method TOPO™-TA
Content And Storage Box 1:
• Topoisomerase I-activated pCR™4-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• T3, T7, M13 forward, and M13 reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid

Store at -68 to -85°C.
Format Kit
For Use With (Application) Chromatin Biology
Product Line One Shot
Product Type Cloning Kit
Promoter T7, T3
Quantity 25 Reactions
Vector TOPO-TA Cloning Vectors

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.