missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, with One Shot™ TOP10 chemically competent E. coli cells

Product Code. 10398572
Change view
Click to view available options
Quantity:
10 Reactions
25 Reactions
50 Reactions
Unit Size:
10 reactions
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10398572 50 Reactions Each
15380204 10 Reactions 10 reactions
10616533 25 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10398572 Supplier Invitrogen™ Supplier No. K460040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Topoisomerase I-activated pCR™II-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; T3, T7, M13 forward, and M13 reverse primers; Control PCR primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid

For fast, efficient cloning and subsequent in vitro transcription/translation

By eliminating time-consuming and tedious restriction site cloning, TOPO cloning is the most reliable method, taking only 5 minutes using a 3-step protocol, and yielding up to 95% recombinants. This kit includes the pCR™II-TOPO TA vector.

  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 and SP6 promoters for efficient in vitro transcription
  • M13 forward and reverse primer sites for sequencing
  • 16 convenient restriction sites, including EcoRI, flanking your insert for subsequent excision or subcloning
  • Kanamycin and ampicillin resistance for your choice of selection in E. coli
  • Easy blue/white colony screening for selection of recombinants
  • Includes OneShot™ TOP10 Competent cells for added convenience and highest cloning efficiencies

Applications:

Cloning, PCR Cloning

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain TOP10
Cell Type Chemically Competent
Cloning Method TOPO™-TA
Content And Storage Box 1:
• Topoisomerase I-activated pCR™II-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid.

Store at -68 to -85°C.
Format Kit
Product Line One Shot
Product Type Cloning Kit
Promoter T7, SP6
Quantity 50 Reactions
Vector TOPO-TA Cloning Vectors

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.