Få mere at vide
Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, with One Shot™ TOP 10 Chemically Competent E.coli, no manual
Produktkode 10422882
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 Reactions
Förpackningsstorlek
Each
Produktkode Quantity unitSize
10422882 50 Reactions Each
1 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 10422882 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' KNM460040
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Denne vare kan ikke returneres. Se returpolitik
For fast, efficient cloning and subsequent in vitro transcription/translation, use Invitrogen's TOPO TA Cloning Dual Promoter Kit with pCR™II vector
By eliminating time-consuming and tedious restriction site cloning, TOPO cloning is the most reliable method, taking only 5 minutes using a 3-step protocol and yielding up to 95% recombinants.
  • 3′-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 and SP6 promoters for efficient in vitro transcription
  • M13 forward and reverse primer sites for sequencing
  • 16 convenient and validated restriction sites including EcoR I flanking your insert for subsequent excision or sub-cloning
  • Kanamycin and ampicillin resistance for your choice of selection in E. coli
  • Easy blue/white colony screening for selection of recombinants
  • Includes OneShot™ TOP10 F′ Competent cells for added convenience and highest cloning efficiency
Cloning, PCR Cloning
TRUSTED_SUSTAINABILITY
Tekniske data
Product Type Kit, Cloning
Includes Box 1: Topoisomerase I-activated pCRII-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid
Bacterial or Yeast Strain TOP10
For Use With (Application) Cloning, PCR Cloning
Quantity 50 Reactions
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.