Learn More
Invitrogen™ ThermoScript™ RT-PCR System for First-Strand cDNA Synthesis
Product Code. 10686723
Click to view available options
:
25 reactions
Product Code. 10686723
Brand: Invitrogen™ 11146024
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Content And Storage 25μL ThermoScript RT (15U/μL); 500μL 5X cDNA Synthesis Buffer; 250μL 0.1M DTT; 100μL 10mM dNTP Mix; 25μL RNaseOUT (40U/μL); 25μL Oligo (dT)20 (50μM); 50μL Random Hexamers (50ng/μL); 1.25mL DEPC-Treated Water; 50μL E. coli RNase H (2U/μL). Store at -20deg.C
Primer Random Primers, Oligo dT Primers
Quantity 25 reactions
For Use With (Application) cDNA Synthesis, RT-PCR