missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ThermoScript™ Reverse Transcriptase
Specifications
Specifications
| Concentration | 15 U/μL |
| Content And Storage | 5X cDNA synthesis buffer (250mM Tris acetate (pH 8.4), 375mM potassium acetate, 40mM magnesium acetate, and stabilizers), and 100mM DTT. Store at -70deg.C |
| Form | Frozen |
| Quantity | 25 reactions (375 units) |
| Sensitivity | Medium |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?