missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ThermoScript™ Reverse Transcriptase

Product Code. 10155132
Change view
Click to view available options
:
100 Reactions; 1500 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10155132 100 Reactions; 1500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10155132 Supplier Invitrogen™ Supplier No. 12236022

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Concentration 15 U/μL
Content And Storage 5X cDNA synthesis buffer (250mM Tris acetate (pH 8.4), 375mM potassium acetate, 40mM magnesium acetate, and stabilizers), and 100mM DTT. Store at -70deg.C
Form Liquid
Format Tube
Quantity 100 reactions (1500 units)
Sensitivity Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.