Learn More
Invitrogen™ ThermalAce™ DNA Polymerase
Extreme thermostability lends versatility
Brand: Invitrogen™ E0200
Code : 50
Additional Details : Weight : 0.02000kg
Includes: ThermalAce DNA Polymerase; 10X reaction buffer; and dNTPs
Description
ThermalAce DNA Polymerase provides high-yield PCR amplification of a variety of templates, especially GC-rich targets, with improved fidelity over TaqPolymerase. The key to the versatility of ThermalAce is its extreme thermostability, which allows it to remain active for long periods at high temperatures (4 hours at >95°C), dramatically improving PCR yields.
- Amplify GC-rich DNA templates (65 to 90% GC content)
- Successfully amplify more of your standard DNA templates
- Achieve higher fidelity than TaqDNA polymerase
Unit Definition:
One unit of ThermalAce DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of dNTPs into acid insoluble material at 74°C.
GC-Rich PCR, PCR, PCR and Real-Time PCR
Specifications
ThermalAce DNA Polymerase; 10X reaction buffer; and dNTPs. Store at -20°C. Guaranteed stable for 6 months when properly stored. | |
Gel Compatibility: Medium | |
200 units | |
Fidelity (vs. Taq): 5X |
For Research Use Only. Not for use in diagnostic procedures.