missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10 Chemically Competent E. coli, no manual
Description
TA Cloning Kits use a pCR vector and yield ≥80% recombinants with ≥90% of the recombinants containing inserts.
- 3′-T overhangs for direct ligation of Taq-amplified PCR products
- Choice of T7 (pCR2.1) or T7 and Sp6 (pCRII) promoters for in vitro RNA transcription and sequencing
- Versatile polylinker with flanking EcoR I sites for easy excision of inserts
- Choice of kanamycin or ampicillin selection
- Easy blue/white colony screening
- M13 forward and reverse primer sites for sequencing
- Manual not included
- Cloning Method: Restriction enzyme/MCS
Cloning, PCR Cloning
Specifications
Specifications
| Bacterial or Yeast Strain | INVαF' |
| Content And Storage | Store One Shot E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
| Includes | Linearized pCR2.1vector, T4 DNA ligase, 10X ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls, One Shot TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Without manual. |
| No. of Reactions | 40 |
| Product Type | One Shot TOP10 Chemically Competent E. coli |
| Quantity | 40 Reactions |
| Vector | TA Cloning Vectors |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?