Learn More
Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10 Chemically Competent E. coli, no manual
Product Code. 10083302
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
Product Code. Quantity unitSize
10083302 20 Reactions Each
1 options
Product Code. 10083302 Supplier Invitrogen™ Supplier No. KNM204001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Bacterial or Yeast Strain TOP10
Content And Storage Store One Shot E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Includes Linearized pCR2.1vector, T4 DNA ligase, 10X ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls, One Shot TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Without manual.
No. of Reactions 20
Product Type One Shot TOP10 Chemically Competent E. coli
Quantity 20 Reactions
Vector TA Cloning Vectors