missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ T-REx™-CHO Cell Line
Specifications
Specifications
| Cell Line | T-REx™-CHO Cell Line |
| Content And Storage | 1 x 107 cells are supplied frozen in 1 ml 90% complete medium and 10% DMSO. Store in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored. |
| System Type | T-Rex™ |
| Species | Hamster |
| Product Line | T-REx |
| Quantity | 3 x 106 Cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?