missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ First-Strand Synthesis System for RT-PCR

Product Code. 10684803
Click to view available options
Quantity:
50 rxns
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10684803

Brand: Invitrogen™ 11904018

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

50μL Oligo(dT)12-18 (0.5μg/μL); 250μL Random hexamers (50ng/μL); 1mL 10X RT buffer; 500μL 25mM Magnesium Chloride; 250μL 0.1M DTT; 250μL 10mM dNTP mix; 50μL SuperScript II RT (50U/μL); 100μL RNaseOUT (40U/μL); 50μL E. coli RNase H (2U/μL); 1.2mL DEPC-treated water; 15μL Control RNA (50ng/μL); 20μL Control Primer A (10μM); 20μL Control Primer B (10μM)

The SuperScript First-Strand Synthesis System for RT-PCR is optimized for the synthesis of first-strand cDNA from purified poly(A)+ or total RNA.

The SuperScript First-Strand Synthesis System for RT-PCR is optimized for the synthesis of first-strand cDNA from purified poly(A)+ or total RNA. The system can be used with as little as 1 ng or as much as 5 μg of total RNA. After synthesis, the cDNA can be amplified with specific primers by PCR without intermediate organic extractions or ethanol precipitations. In conjunction with PCR, the system can be used to detect the presence of rare messages, to quantitate the amount of specific mRNA from small numbers of cells, or to clone specific cDNAs without constructing an entire cDNA library. The system is flexible, allowing the use of any PCR enzyme. Combine with AccuPrime Taq DNA Polymerase or Platinum Taq DNA Polymerase for higher specificity PCR or with AccuPrime Pfx DNA Polymerase for high-fidelity cloning applications.

SuperScript II RT
The first-strand cDNA synthesis reaction is catalyzed by SuperScript II Reverse Transcriptase (RT). This enzyme has been engineered to reduce the RNase H activity that degrades mRNA during the first-strand reaction, resulting in greater full-length cDNA synthesis and higher yields of first-strand cDNA than obtained with RNase H+ RTs. Because SuperScript II RT is not inhibited significantly by ribosomal and transfer RNA, it may be used effectively to synthesize first-strand cDNA from a total RNA preparation. The enzyme exhibits increased thermal stability and may be used at temperatures up to 50°C.

Using the SuperScript First-Strand Synthesis System for RT-PCR
This system has been optimized to synthesize first-strand cDNA from varying amounts of starting material. The SuperScript II RT concentration has been lowered and RNaseOUT Recombinant RNase Inhibitor has been added to the system as part of this optimization process. Additionally, reaction conditions have been modified to further increase the sensitivity of the system. Using the kit, you synthesize first-strand cDNA using either total RNA or poly(A)+ selected RNA primed with oligo(dT), random primers, or a gene-specific primer. Then perform PCR in a separate tube using primers specific for the gene of interest.

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• Oligo(dT)12-18, 50 μL (0.5 μg/μL)
• Random hexamers, 250 μL (50 ng/μL)
• 10X RT buffer, 1 mL
• Magnesium chloride, 500 μL (25 mM)
• DTT, 250 μL (0.1 M)
• dNTP mix, 250 μL (10 mM)
• SuperScript™ II RT, 50 μL (50 U/μL)
• RNaseOUT™, 100 μL (40 U/μL)
E. coli RNase H, 50 μL (2 U/μL)
• DEPC-treated water, 1.2 mL
• Control RNA, 15 μL (50 ng/μL)
• Control Primer A, 20 μL (10 μM)
• Control Primer B, 20 μL (10 μM)

Store at –20°C.

Format Kit
GC-Rich PCR Performance High
Reaction Speed 50 min.
Technique Reverse Transcription
Optimal Reaction Temperature 42°C
Primer Random Primers, Oligo dT Primers
Quantity 50 rxns
Reverse Transcriptase SuperScript II
Shipping Condition Wet Ice
For Use With (Application) RT-PCR
Final Product Type First-Strand cDNA
No. of Reactions 50 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 12 kb
Starting Material RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.