missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat miR RNAi Select

Product Code. 10638493
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10638493 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10638493 Supplier Invitrogen™ Supplier No. 100002320
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Easily design effective RNAi molecules for reliable results

  • Using BLOCK-iT™ RNAi Designer, design Custom BLOCK-iT siRNA sequences for efficient and specific gene expression knockdowns in any organism
  • For enhanced target specificity and stability in serum, Stealth RNAi™ RNAi chemistry is available

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
RNAi Type siRNA
Product Type Rat miR RNAi Select
Quantity 1 Each
Target Rat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.