missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Quant-iT™ PicoGreen™ dsDNA Assay Kits and dsDNA Reagents

Product Code. 10398702
Change view
Click to view available options
Quantity:
1 mL
1 mL kit
10 x 100 μL
10 x 100 μL kit
Product Type:
dsDNA Assay Kit
dsDNA Reagent
Packaging Type:
1 x 1 mL Bottle
10 x 100 μL Tubes
Unit Size:
1000µL
1mL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Packaging Type unitSize
10398702 10 x 100 μL kit dsDNA Assay Kit 10 x 100 μL Tubes Each
10545213 1 mL kit dsDNA Assay Kit 1 x 1 mL Bottle 1mL
10535213 1 mL dsDNA Reagent 1 x 1 mL Bottle 1mL
11559196 10 x 100 μL dsDNA Reagent 10 x 100 μL Tubes 1000µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10398702 Supplier Invitrogen™ Supplier No. P11496

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Selectively detect dsDNA from a wide variety of sources, and even in presence of ssDNA, RNA, and free nucleotides, with the Quant-iT PicoGreen dsDNA Assay Kit and PicoGreen dsDNA Reagent.

Achieve precise and accurate dsDNA concentration measurements over a broad dynamic range with the Quant-iT Picogreen dsDNA Assay kits and dsDNA reagents. Picogreen assays are compatible with most microplate readers and fluorometers.

Rapidly detect as little as 0.25 pg/uL of dsDNA, even in the presence of ssDNA, RNA, and free nucleotides, using the PicoGreen dsDNA quantitation assay and reagent. The assay is linear over three orders of magnitude and has little sequence dependence, allowing you to accurately measure DNA from many sources, including genomic DNA, viral DNA, miniprep DNA, or PCR amplification products.

PicoGreen dsDNA quantitation reagents are
• Significantly more sensitive than UV absorbance readings, saving precious sample amounts
• Specific for dsDNA in the presence of RNA
• Easy to use—just add the dye working solution to the sample, incubate five minutes, and read
• Suitable for use in 96- and 384-well plate formats
• Compatible with most fluorescence-based microplate readers and fluorometers
• Picogreen aproximate fluorescence absorption/emission maxima is 502/523 nm, bound to dsDNA

Applications
The PicoGreen dsDNA quantitation reagent and kits are ideal for PCR-based assays, microarray samples, DNA damage assays, enzyme activity assays, genomic DNA quantitation, measuring dsDNA in complex mixtures, and viral DNA quantitation.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Excitation/Emission 500/525
For Use With (Equipment) Microplate Reader
No. of Reactions 200 Assays (2 mL assay volume)
Packaging Type 10 x 100 μL Tubes
Quantitation Range 50 pg to 2 μg
Product Line PICOGREEN, Quant-iT
Product Type dsDNA Assay Kit
Quantity 10 x 100 μL kit
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.