missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Quick Gel Extraction Kit
GREENER_CHOICE

Product Code. p-4926495
Click to view available options
Quantity:
250 Preps
50 Preps
Unit Size:
1 set
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10749243

Brand: Invitrogen™ K210012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Gel Solubilization Buffer (L3)
Wash Buffer (W1)
Elution Buffer (E5); (10 mM Tris-HCl, pH 8.5)
Quick Gel Extraction Columns
Wash Tubes
Recovery Tubes

The PureLink™ Quick Gel Extraction Kit allows you to rapidly and efficiently purify DNA fragments from TAE or TBE agarose gels of various percentages.

The PureLink™ Quick Gel Extraction Kit allows you to rapidly and efficiently purify DNA fragments from TAE or TBE agarose gels of various percentages. DNA can be extracted and purified from agarose gels with different melting points in ∼30 minutes using PureLink™ silica membrane-based quick gel extraction columns. For your convenience, purification protocols are provided for centrifugation and for vacuum.

Advantages of using PureLink™ Quick Gel Extraction Kit:

  • Purify DNA fragments from TAE and TBE agarose gels of various percentages and melting points
  • Complete the procedure in ∼30 minutes
  • Easily purify DNA fragments from 40 bp to 10 kb from gels
  • Obtain high recovery of DNA fragments
  • Bind and purify up to 15 μg DNA with one column
  • Purify DNA fragments that are high quality and show reliable performance in PCR, restriction enzyme digestion, cloning, and labeling

Note: The PureLink™ Quick Gel Extraction Kit is not designed to purify supercoiled plasmid DNA or genomic DNA from agarose gels. Only linear DNA fragments may be purified from gels using this kit.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage PureLink Elution Tubes
For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
Isolation Technology Silica Spin Column
Sample Type Agarose Gels, Gel Samples
Elution Volume 50 μL
Final Product Type DNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
Green Features Less hazardous, sustainable packaging
High-throughput Compatibility Not High-throughput Compatible (Manual)
Label or Dye Ethidium Bromide, SYBR Safe
Quantity 50 Preps
Shipping Condition Room Temperature
Test Time 30 min.
Yield 15 μg (Binding capacity)
Starting Material Amount ≤400 mg
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 0.12 LT PURELINK GEL EXTRACTION KIT
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Thiocyanic acid, compound with guanidine

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.