missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ HiPure Precipitator Module

Product Code. 10105393
Change view
Click to view available options
Quantity:
10 Preps
25 Preps
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10105393 10 Preps Each
10294023 25 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10105393 Supplier Invitrogen™ Supplier No. K210021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

The PureLink HiPure Precipitator Module includes HiPure Precipitators, and 5mL and 30mL syringes.

The PureLink™ HiPure Precipitator allows fast, simple, and efficient desalting and concentration of plasmid DNA isolated using anion-exchange chromatography.

  • Allows fast, simple, and efficient desalting and concentration of plasmid DNA isolated using anion-exchange chromatography
  • Traditional method for DNA precipitation of anion exchange isolated plasmid DNA involves isopropanol precipitation followed by centrifugation and drying of DNA which is time consuming and labor intensive
  • Allows plasmid DNA concentration within 5 minutes, eliminates need for centrifugation, and reduces risk of losing DNA pellet during supernatant removal, and recovery is >80%

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The PureLink™ HiPure Precipitator Module includes HiPure Precipitators, and 5 mL and 30 mL syringes. Store all components at room temperature.
Format Syringe
Isolation Technology Anion Exchange Resin
Sample Type Bacterial Culture
Final Product Type Plasmid DNA
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility Not High-throughput Compatible (Manual)
Prep Scale > 200 μg (Large-Scale) Plasmid DNA
Product Line PureLink
Product Type Precipitator Module
Quantity 10 Preps
Shipping Condition Room Temperature
Target Plasmid DNA
Test Time 5 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.