missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ HiPure Plasmid Gigaprep Kit

Product Code. 10033432
Change view
Click to view available options
:
2 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10033432 2 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10033432 Supplier Invitrogen™ Supplier No. K210009

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

RNase A (20 mg/mL in R3), Lysis Buffer (L7), Precipitation Buffer (N3), Equilibration Buffer (EQ1), Wash Buffer (W8), Elution Buffer (E4), TE Buffer (TE) Columns

No phenol, chloroform, ethidium bromide, and cesium chloride

  • Fast, simple protocols - Purify your plasmid DNA in less than two hours
  • Labor-saving - No need for extra steps to remove endotoxin and other contaminants
  • Megapreps and Gigapreps save time by utilizing vacuum-assisted cartridges
  • PureLink HiPure Plasmid Purification Systems are designed to isolate plasmid DNA of the highest purity (1) with wide scale range
  • Simple protocols an anion-exchange resin allow you to purify plasmid DNA at quality equivalent to two passes through cesium chloride gradients - the most rigorous method for DNA purification
  • In less than two hours, DNA is pure enough for transfections with no need for additional steps to remove contaminants like RNA, proteins, and endotoxin
  • Additionally, exposure to and disposal of hazardous materials is minimized

DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Plasmid DNA Purification

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Isolation Technology Anion Exchange Resin
Sample Type Bacterial Culture
For Use With (Application) Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
No. of Reactions 2 Preps
Sample Volume >200μg (Large-Scale) Plasmid DNA
Target Plasmid DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.