missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PureLink™ Foil Tape
Click to view available options
Quantity:
50 Pieces
Unit Size:
Pack of 50
Description
Includes
Package of 50 pieces
PureLink Foil Tape offers a unique approach to high throughput plasmid DNA isolation in the PureLink 96 Plasmid Purification System.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Silver |
| Content And Storage | Package of 50 pieces The PureLink™ 96 Components are shipped at room temperature. Upon receipt, store the components at room temperature. |
| For Use With (Application) | Automated Nucleic Acid Purification, DNA Extraction |
| Plate Compatibility | 384-well Plates, 96-well Plates |
| Product Line | PureLink |
| Product Type | Foil Tape |
| Quantity | 50 Pieces |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction