missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Air Porous Tape

Product Code. 10544453
Change view
Click to view available options
Quantity:
50 Pieces
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10544453 50 Pieces Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10544453 Supplier Invitrogen™ Supplier No. 12262010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Package of 50 pieces

PureLink Air Porous Tape offers method for high throughput plasmid DNA isolation in PureLink 96 Plasmid Purification System and allows for proper aeration of bacterial cultures.

PureLink Air Porous Tape offers method for high throughput plasmid DNA isolation in PureLink 96 Plasmid Purification System and allows for proper aeration of bacterial cultures.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage Package of 50 pieces The PureLink™ 96 Components are shipped at room temperature. Upon receipt, store the components at room temperature.
For Use With (Application) Automated Nucleic Acid Purification, DNA Extraction
Plate Compatibility 384-well Plates, 96-well Plates
Product Line PureLink
Product Type Air Porous Tape
Quantity 50 Pieces
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.