Learn More
Invitrogen™ pTrcHis TOPO™ TA Expression Kit
Product Code. 10358392
Change view
Click to view available options
Quantity:
20 reactions
Unit Size:
Each
Product Code. Quantity unitSize
10358392 20 reactions Each
1 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 10358392 Supplier Invitrogen™ Supplier No. K441001
For Research Use Only. All usage must comply with product instructions.
Includes
The pTrcHis-2-TOPO TA Expression Kit contains two boxes. The TOPO TA box contains 200ng of linearized, topoisomerase I-activated pTrcHis-TOPO or pTrcHis2-TOPO vector, sterile water, dNTPs, 10X PCR buffer, a control template and primers, primers for sequencing or PCR screening, and an expression control plasmid. The TOP10 box contains 21 x 50μL aliquots of chemically competent TOP10 E. coli, S.O.C. medium, and a supercoiled control plasmid.
Allow fast, efficient cloning of PCR products into prokaryotic expression vector for high-level, regulated expression from trc promoter
  • Vector includes minicistron element for enhanced translation efficiency of eukaryotic proteins in E. coli
  • Vector provided linearized and activated with topoisomerase I for 5-minute TOPO Cloning with >85% cloning efficiency
  • N-terminal Xpress epitope for detection with an Anti-Xpress Antibody
  • N-terminal polyhistidine (6xHis) tag for purification using nickel-chelating resin and detection with Anti-HisG Antibody
  • Enterokinase cleavage site for efficient removal of N-terminal fusion tags
  • C-terminal polyhistidine (6xHis) tag for purification using nickel-chelating resin and detection with Anti-His(C-term) Antibody
Bacterial Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Constitutive or Inducible System Inducible
Inducing Agent IPTG
Promoter Trc, lacO
Product Type TA Expression Kit
Selection Agent (Eukaryotic) None
Content And Storage The pTrcHis- and pTrcHis2-TOPO™ TA Expression Kits each contain two boxes. The TOPO™ TA box contains 200 ng of linearized, topoisomerase I-activated pTrcHis-TOPO™ or pTrcHis2-TOPO™ vector, sterile water, dNTPs, 10X PCR buffer, a control template and primers, primers for sequencing or PCR screening, and an expression control plasmid. The One Shot™ TOP10 box contains twenty-one, 50-μl aliquots of chemically competent TOP10 E. coli, S.O.C. medium, and a supercoiled control plasmid. Store the TOPO TA Cloning™ box at -20°C. Store the One Shot™ box at -80°C. All reagents are guaranteed stable for 6 months when properly stored.
Antibiotic Resistance Bacterial Ampicillin (AmpR)
Bacterial or Yeast Strain TOP10
Cleavage EK (Enterokinase) Recognition Site
Expression System E. coli
Expression Mechanism Cell-Based Expression
Protein Tag His Tag (6x), Xpress Epitope Tag
Cloning Method TOPO-TA
Quantity 20 reactions
Vector pTrc
Product Line TOPO