Learn More
Invitrogen™ pRSET-BFP Bacterial Expression Vector
Product Code. 11550826
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
For Research Use Only. All usage must comply with product instructions.
Product Code. 11550826
Brand: Invitrogen™ V35420
For Research Use Only. All usage must comply with product instructions.
Allow fusion of a protein of interest to the widely used and well-characterized fluorescent proteins from the jellyfish Aequorea victoria
  • These vectors contain the EGFP, for Emerald Green Fluorescent Protein (EmGFP), which has been further optimized for mammalian expression
  • Vector also codes for Cyan (CFP) and Blue (BFP) Fluorescent Proteins for simple, noninvasive detection of recombinant protein
Bacterial Expression, Enzyme and Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Protein Expression, Proteins, Expression, Isolation and Analysis, Reporter Assays
TRUSTED_SUSTAINABILITY
Specifications
Constitutive or Inducible System Inducible
Inducing Agent IPTG
Promoter T7
Product Type Bacterial Expression Vector
Reporter Gene BFP
Selection Agent (Eukaryotic) None
Content And Storage Store in freezer (-5 to -30°C).
Antibiotic Resistance Bacterial Ampicillin (AmpR)
Cleavage EK (Enterokinase) Recognition Site
Protein Tag His Tag (6x), Xpress Epitope Tag
Cloning Method Restriction Enzyme/MCS
Quantity 10 μg
Vector pRSET
Product Line Vivid Colors