missing translation for 'onlineSavingsMsg'
Learn More

SuperScript™, Platinum™, SYBR™ SuperScript™ III Platinum™ SYBR™ Green One-Step qPCR Kit w/ROX

Product Code. p-4922118
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11588656

Brand: SuperScript™, Platinum™, SYBR™ 11746100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Combine a powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.

One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient, high-throughput one-step format. SuperScript™ III Reverse Transcriptase produces higher yields of full-length cDNA for broader gene representation. Platinum™ Taq DNA Polymerase activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification. ROX reference dye premixed at optimal concentration for ABI instruments.

  • Provides a highly efficient, cost-effective qRT-PCR system with SYBR Green I detection

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Store in freezer (-5 to -30°C).
Detection Method SYBR
PCR Method 1-step RT-qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT System
Product Line Platinum, SYBR, SuperScript
Product Type Real Time PCR SYBR Master Mix
Quantity 100 reactions
Shipping Condition Dry Ice
Sufficient For 100 Reactions
For Use With (Application) Gene Expression
No. of Reactions 100 Reactions
Sample Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.