missing translation for 'onlineSavingsMsg'
Learn More

Platinum™, SYBR™ GreenER™ SYBR GreenER™ qPCR SuperMix Universal

Product Code. 10432153
Change view
Click to view available options
No. of Reactions:
100 Reactions
2000 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10432153 500 Reactions 500 reactions
10543533 2000 Reactions Each
10553533 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10432153 Supplier Platinum™, SYBR™ GreenER™ Supplier No. 11762500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

2X SYBR GreenER qPCR SuperMix and a separate tube of ROX reference dye

Incorporate the latest high-performance technology to provide the most reliable gene expression data on a variety of platforms including ABI 7500, Corbett Rotor-Gene™ etc.

SYBR™ GreenER™ qPCR SuperMixes Universal incorporate the latest high-performance technology to provide the most reliable gene expression data on a variety of platforms, including ABI 7500, Corbett Rotor-Gene™, MJ Chromo4™ and Opticon™, and Stratagene instruments. The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility with. A novel dsDNA-binding dye exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings. An optimized buffer system improves sensitivity and provides excellent long-term stability. Uracil DNA glycosylase reduces carryover contamination in qPCR.

Eight cDNA targets and eight genomic DNA targets were amplified from 1ng of cDNA and 6pg of gDNA, respectively, on the ABI PRISM™ 7900HT Sequence Detection System using the SYBR™ GreenER™ qPCR SuperMix for ABI PRISM™ instruments. For all 16 targets, detection was positive in the presence of template but negative for all no-template controls.

  • The SYBR™ GreenER™ qPCR SuperMix kits include a new dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range

Consistent Specificity and Low-Copy Detection:
  • The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets
  • The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity
  • With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains SYBR™ GreenER™ qPCR SuperMix Universal (12.5 mL). SYBR™ GreenER™ qPCR SuperMix Universal is a 2X SuperMix with ROX Reference dye provided in a separate tube (500 μL). Sufficient reagents are provided for 500 reactions based on a 50 μL reaction size.

Store at +4°C upon receipt.
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) 7500 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P, MJ Chromo4, MJ Opticon
Product Line Platinum, SYBR GreenER
Product Type Real Time PCR SYBR Master Mix
Quantity 500 reactions
Shipping Condition Dry Ice
Sufficient For 500 Reactions of 50 μL
For Use With (Application) Gene Expression
No. of Reactions 500 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.