missing translation for 'onlineSavingsMsg'
Learn More

Platinum™, SYBR™ Platinum™ SYBR™ Green qPCR SuperMix-UDG w/ROX

Product Code. 11548656
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11548656

Brand: Platinum™, SYBR™ 11744100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

2X SuperMix with SYBR Green I and ROX Reference Dyes, and 50mM MgCl2

Ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA.

Platinum™ Taq DNA Polymerase activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification. Uracil-DNA Glycosylase (UDG) prevents carryover contamination in conjunction with dUTP (included) for improved specificity. ROX reference dye is included in the mix for maximum convenience. The ROX reference dye premixed at optimal concentration for ABI instruments.

Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX provides a sensitive, cost-effective system for real-time PCR detection with SYBR™ Green I dye

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Store in freezer (-5 to -30°C).
Detection Method SYBR
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System
Product Line Platinum, SYBR
Product Type Real Time PCR SYBR Master Mix
Quantity 100 reactions
Shipping Condition Dry Ice
Sufficient For 100 Reactions
For Use With (Application) Gene Expression
Compatibility ABI PRISM™ 7000, ABI PRISM™ 7700, ABI PRISM™ 7900HT, ABI PRISM™ 7900HT Fast, ABI 7300 Real-Time PCR System, ABI GeneAmp™ 5700
No. of Reactions 100 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.