Learn More
Platinum™, SYBR™ Platinum™ SYBR™ Green qPCR SuperMix-UDG w/ROX
Description
Includes
2X SuperMix with SYBR Green I and ROX Reference Dyes, and 50mM MgCl2
Platinum™ Taq DNA Polymerase activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification. Uracil-DNA Glycosylase (UDG) prevents carryover contamination in conjunction with dUTP (included) for improved specificity. ROX reference dye is included in the mix for maximum convenience. The ROX reference dye premixed at optimal concentration for ABI instruments.
Order Info
Shipping Conditions: Dry Ice
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | Contains 2X Platinum SYBR Green qPCR SuperMix-UDG with ROX (12.5 mL) and 50 mM Magnesium Chloride (2 × 1 mL). Contains sufficient reagents for 500 reactions of 50 μL each. Store in freezer (-5 to -30°C). |
| Detection Method | SYBR |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System |
| Product Line | Platinum, SYBR |
| Product Type | Real Time PCR SYBR Master Mix |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.