missing translation for 'onlineSavingsMsg'
Learn More

SuperScript™, Platinum™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit w/ROX

Product Code. 11568656
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
11568656 100 Reactions 100 reactions
11578656 500 Reactions 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11568656 Supplier SuperScript™, Platinum™ Supplier No. 11745100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

SuperScript™ III/Platinum™ Taq Mix, 2X Reaction Mix with ROX and 1mL Magnesium Sulfate (50mM). Guaranteed stable for 6 months when properly stored.

One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.

One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient, high-throughput one-step format. One-Step qRT-PCR kits with ROX include SuperScript™ III Reverse Transcriptase which produces higher yields of full-length cDNA for broader gene representation, Platinum™ Taq DNA Polymerase which activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification and ROX reference dye premixed at optimal concentration for ABI instruments

Two kit configurations allow your choice of detection methods:
  • SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit provides a highly efficient, cost-effective qRT-PCR system with SYBR Green I detection
  • SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection for use with LUX™ fluorogenic primers or dual-labeled probes (e.g. TaqMan™)

Order Info

Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 100 μL SuperScript™ III/Platinum™ Taq Mix
• 2 × 1.25 mL 2X Reaction Mix with ROX
• 1 mL Magnesium Sulfate (50 mM)

Store components in the dark at -20°C.
Detection Method Primer-probe
PCR Method 1-step RT-qPCR
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT System
Product Line Platinum, SuperScript
Product Type One-Step qRT-PCR Kit with Rox
Quantity 100 reactions
Shipping Condition Dry Ice
Sufficient For 100 Reactions
For Use With (Application) Gene Expression, Pathogen Detection, Virus Detection
No. of Reactions 100 Reactions
Sample Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.