missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ PCR SuperMix 96, Non-skirted

Product Code. 10614623
Click to view available options
:
480 reactions
This item is not returnable. View return policy

Product Code. 10614623

missing translation for 'mfr': Invitrogen 11306073

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Each 22.5μL volume of Platinum PCR SuperMix 96 is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solution. Reagents are provided for 5 x 96 amplification reactions of 25μL each. 480 rxns: 5 nonskirted plates containing 22.5μL of Platinum PCR SuperMix. 480 reactions.

Prealiquoted reagents simplify high-throughput applications

Platinum PCR SuperMix 96 provides qualified reagents for the amplification of nucleic acid templates by polymerase chain reaction (PCR). The reagents are prealiquoted into 96-well microtiter plates (nonskirted) for high-throughput applications as well as general ease of use. Each plate well contains anti-Taq DNA polymerase antibody, Mg++, deoxyribonucleotide triphosphates, and recombinant Taq DNA Polymerase. Due to specific binding of the anti-Taq antibody, polymerase activity is inhibited at room temperature. Activity is restored after a denaturation step in PCR cycling at 94°C, providing an automatic hot start in PCR and thereby improving PCR specificity and yield.

Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR

Order Info

Shipping condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each 22.5μL volume of Platinum PCR SuperMix 96 is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solution. Reagents are provided for 5 x 96 amplification reactions of 25μL each.

480 rxns: 5 nonskirted plates containing 22.5μL of Platinum PCR SuperMix
Quantity 480 reactions
For Use With (Application) RT-PCR
No. of Reactions 480
Overhang 3'-A
Size (Final Product) 5kb or less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.