missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Platinum™ PCR SuperMix
Description
The mixture contains anti-Taq DNA polymerase antibody, Mg++, dNTPs, and recombinant Taq DNA polymerase at concentrations sufficient to allow amplification during PCR. Platinum™ PCR SuperMix is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 100 or 5000 amplification reactions of 50 μL each are provided.
Specifications
Specifications
| Content And Storage | Platinum PCR SuperMix is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 5000 amplification reactions of 50μL each are provided. Platinum PCR SuperMix Components: 22U/mL complexed recombinant Taq DNA polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM MgCl2, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, and stabilizers. 100 rxns: 4 x 1.125mL |
| Quantity | 100 reactions |
| For Use With (Application) | Real Time PCR (qPCR), Standard PCR |
| No. of Reactions | 100 |
| Overhang | 3'-A |
| Size (Final Product) | 5kb or less |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?