missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Platinum™ PCR SuperMix
Click to view available options
:
5000 reactions
Specifications
Specifications
| Content And Storage | Platinum PCR SuperMix is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 5000 amplification reactions of 50μL each are provided. Platinum PCR SuperMix Components: 22U/mL complexed recombinant Taq DNA polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM MgCl2, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, and stabilizers. 5000 rxns: 4 x 56.25mL |
| Quantity | 5000 reactions |
| For Use With (Application) | RT-PCR |
| No. of Reactions | 5000 |
| Overhang | 3'-A |
| Size (Final Product) | 5kb or less |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction