Learn More
Invitrogen™ Platinum™ Pfx DNA Polymerase
Description
Includes
Platinum Pfx DNA Polymerase (100 units total, at 2.5U/μL) is a supplied with a vial (1mL) of 10X PCRx Enhancer solution, a vial (1mL) of 10X Pfx amplification buffer, and a vial (1mL) of 50mM MgSO4. Designed for 100 reactions (at 1 unit each in a 50μL reaction volume).
Platinum Pfx DNA Polymerase is ideal for amplification of DNA fragments for high-fidelity PCR applications. High fidelity is provided by a proprietary enzyme preparation containing recombinant DNA polymerase from Thermococcusspecies KODwith proofreading (3'-5' exonuclease) activity. Platinumantibody technology provides a simple, automatic, hot-start method that improves PCR specificity. PCRx Enhancer Solution is included for higher primer specificity, broader magnesium concentration allowances, broader annealing temperature parameters, and improved thermostability of PlatinumPfxDNA Polymerase. The PCRx Enhancer Solution also helps optimize PCR of problematic and/or GC-rich templates.
• 26 times higher fidelity than Taq DNA polymerase
• Amplification of fragments up to 12 kb
• Room temperature reaction assembly
Applications
Amplification of DNA from complex genomic, viral, and plasmid templates; and RT-PCR. Platinum Pfx DNA Polymerase is especially effective when very high levels of fidelity are required.
Unit definition
One unit incorporates 10 nmoles of deoxyribonucleotide into acid-precipitable material in 30 minutes at 74°C.
Order Info
Approved for shipment on Wet or Dry Ice
Specifications
Specifications
| Quantity | 100 Reactions |
| Includes | Platinum Pfx DNA Polymerase (100 units total, at 2.5 U/μL) is a supplied with: • 10X PCRx Enhancer solution, 1 mL • 10X Pfx amplification buffer, 1 mL • 50 mM MgSO4, 1 mL Designed for 100 reactions (at 1 unit each in a 50 μL reaction volume) Store at -20°C. |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.