missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Blue PCR SuperMix

Product Code. 10144992
Change view
Click to view available options
1000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10144992 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10144992 Supplier Invitrogen™ Supplier No. 12580023

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Content And Storage 4 x 11.25mL Composition: Each tube contains 22U/mL complexed recombinant Taq DNA Polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM magnesium chloride, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, stabilizers, glycerol, and blue tracking dye.
Quantity 1000 reactions
For Use With (Application) Standard PCR
Overhang 3'-A
Size (Final Product) 5kb or less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.