missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Blue PCR SuperMix 96, Skirted

Product Code. 11538716
Change view
Click to view available options
5 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11538716 5 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11538716 Supplier Invitrogen Supplier No. 12580031

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Platinum Taq is supplied with 10X PCR buffer (200mM Tris-HCl (pH 8.4), 500mM KCl), and 50mM MgCl2. Platinum Taq PCRx DNA Polymerase is supplied with 10X PCR buffer, 50mM MgCl2 , 50mM MgSO4 , 10X PCRx Enhancer Solution, and 10X PCRx Amplification Buffer. SuperMixes are supplied in a 1.1X concentration and contain Platinum Taq, dNTPs, MgSO4 ,and buffer. Platinum Blue PCR SuperMix also contains loading/tracking dye.

Ideal for automatic hot-start amplification of DNA fragments with improved specificity over Taq DNA polymerase

Derived from recombinant Taq DNA polymerase by binding of a thermolabile inhibitor containing monoclonal antibodies to Taq DNA polymerase. During initial denaturation step of PCR, the inhibitor is denatured and active Taq DNA polymerase is released into the reaction. Platinum Blue PCR SuperMix is a PCR master mix containing Platinum Taq DNA Polymerase, dNTPs, premixed loading/tracking dye, buffer, magnesium and salts. The blue tracking buffer will not inhibit your PCR performance. After PCR cycling, simply load your PCR product directly onto an agarose gel. Platinum TaqPCRx DNA Polymerase is Platinum Taq DNA Polymerase supplied with the PCRx Enhancer System for optimizing PCR of problematic and/or GC-rich templates. 10X PCRx Enhancer Solution with 10X PCRx Amplification Buffer offers higher primer specificity, broader magnesium concentration, broader annealing temperature, and improved thermostability of Taq DNA polymerase.

  • Improved specificity and yields of PCR compared to Taq DNA polymerase without “hot-start”
  • PCR products up to 5kb in size
  • Room temperature reaction assembly
Applications: Amplification of DNA from complex genomic, viral, and plasmid templates; and RT-PCR.
Unit Definition: One unit of Platinum TaqDNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of deoxyribonu-cleotide into DNA in 30 min. at 74°C.

Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR

Order Info

Shipping condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Platinum Taq is supplied with 10X PCR buffer (200mM Tris-HCl (pH 8.4), 500mM KCl), and 50mM MgCl2 . Platinum TaqPCRx DNA Polymerase is supplied with 10X PCR buffer, 50mM MgCl2 , 50mM MgSO4 , 10X PCRx Enhancer Solution, and 10X PCRx Amplification Buffer. SuperMixes are supplied in a 1.1X concentration and contain Platinum Taq, dNTPs, MgSO4 ,and buffer. Platinum Blue PCR SuperMix also contains loading/tracking dye.
Quantity 5 plates
For Use With (Application) Real Time PCR (qPCR)
Overhang 3'-A
Size (Final Product) 5kb or less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.