missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Blue PCR SuperMix

Product Code. 10442862
Click to view available options
:
100 reactions
This item is not returnable. View return policy

Product Code. 10442862

Brand: Invitrogen™ 12580015

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Premixed reagents for high-throughput applications

Platinum Blue PCR SuperMix provides qualified reagents for the amplification of nucleic acid templates by PCR, and includes blue tracking dye for subsequent gel analysis. The reagents are premixed for high-throughput applications as well as general ease of use.

Using Platinum Blue PCR SuperMix
The mixture contains recombinant Platinum Taq DNA polymerase, Mg++, dNTPs, glycerol (used as a precipitant), and blue tracking dye. Platinum Blue PCR SuperMix is supplied at a 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents are provided sufficient for 100 or 1,000 amplification reactions of approx. 50μL each.

Hot-start specificity
Specific binding of the anti-Taq antibody inhibits polymerase activity at room temperature. Activity is restored after a denaturation step in PCR cycling at 94°C, providing an automatic “hot start” in PCR and improving specificity and yield.

Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4 x 1.125mL Composition: Each tube contains 22U/mL complexed recombinant Taq DNA Polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM magnesium chloride, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, stabilizers, glycerol, and blue tracking dye.
Quantity 100 reactions
For Use With (Application) Real Time PCR (qPCR), Standard PCR
No. of Reactions 100
Overhang 3'-A
Size (Final Product) 5kb or less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.