missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Plasmid pCMV·SPORT-βgal

Product Code. 11563127
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11563127 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 11563127 Supplier Invitrogen™ Supplier No. 10586014
  • ONLINE EXCLUSIVE PRICE
    £329.00
    £246.75 / 25µg
    Save £82.25
    25% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Used as a positive control for monitoring expression in eukaryotic cells

  • Plasmid contains reporter gene b-galactosidase (b-gal) from E. coli cloned as Not I fragment into plasmid pCMV·SPORT1
  • Plasmid also contains CMV promoter upstream of b-gal gene, followed by the SV40 t-intron and polyadenylation signal
  • b-lactamase gene allows selection for ampicillin resistance in E. coli

Antibody and Cell Culture Production, Beta-Galactosidase Assays, Bioproduction, Constitutive Expression/Enzyme and Protein Activity Assays, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Transfection
Promoter CMV
Product Type Plasmid
Reporter Gene Beta-Gal (lacZ)
Selection Agent (Eukaryotic) None
Content And Storage Store at +4°C. Store at -20°C long term.
Shipping Condition Approved for shipment on Wet or Dry Ice
Protein Tag Untagged
Quantity 25 μg
Vector pCMV
For Use With (Application) Reporter Assays, Constitutive Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.