missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pDisplay™ Mammalian Expression Vector
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Includes
20μg pDisplay vector is supplied supercoiled and lyophilized.
- Displayed proteins can be analyzed for their ability to interact with known or putative ligands
- Proteins of interest are targeted and anchored to cell surface by cloning the gene of interest in frame with vectors unique N-terminal secretion signal and C-terminal transmembrane anchoring domain of platelet-derived growth factor receptor (PDGFR)
- In contrast to phage display vectors which operate exclusively in prokaryotic cells, pDisplay vector offers advantage of having displayed protein of interest processed in mammalian cells
- Therefore, recombinant proteins of eukaryotic origin that are expressed from pDisplay more closely resemble their native form: this may favor more accurate ligand binding interactions
- In addition to N-terminal cell surface targeting signal and C-terminal transmembrane anchoring domain, pDisplay vector contains T7 promoter/priming site for in vitro transcription of sense RNA and for sequencing of inserts
- Neomycin resistance marker for stable selection in mammalian cells using Geneticin™
- SV40 origin for replication and simple vector rescue in cell lines expressing large T antigen (e.g.,COS-1 and COS-7)
- Ampicillin resistance gene for selection in E. coli
Mammalian Two-Hybrid, Protein-Protein Interactions, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Delivery Type | Transfection |
| Promoter | CMV |
| Product Type | Mammalian Expression Vector |
| Selection Agent (Eukaryotic) | Geneticin™ (G-418) |
| Content And Storage | 20 μg pDisplay vector is supercoiled and supplied as a 0.5 μg/mL solution in TE buffer, pH 8.0. Store at -20°C. Guaranteed stable for 6 months when properly stored. |
| Protein Tag | IgK Leader Sequence, PDGFR Transmembrane Domain, c-Myc Epitope Tag, HA Tag |
| Cloning Method | Restriction Enzyme/MCS |
| Quantity | 20 μg |
| Vector | pDisplay |
| Product Line | pDISPLAY |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction