missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pcDNA™3.1/V5-His TOPO™ TA Expression Kit
Provides efficient, 5-minute, one-step cloning strategy for direct insertion of Taq polymerase-amplified PCR products into plasmid vector
£742.00 - £1384.00
Specifications
Constitutive or Inducible System | Constitutive |
---|---|
Delivery Type | Transfection |
Promoter | CMV |
Product Type | TOPO TA Expression Kit |
Selection Agent (Eukaryotic) | Geneticin™ (G-418) |
Product Code | Brand | Quantity | Vector | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Vector | Price | Quantity & Availability | ||||
10575383
|
Invitrogen™
K480001 |
20 Reactions | TOPO-TA Vectors, pcDNA |
£742.00
Each |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10584043
|
Invitrogen™
K480040 |
40 Reactions | TOPO-TA Vectors, pcDNA |
£1384.00
Each |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- No ligase, post-PCR procedures, or PCR primers containing specific sequences are required
- Once cloned, analyzed, and transfected, PCR product will express directly in mammalian cell lines
- pcDNA3.1/V5-His TOPO vector features strong CMV promoter for high-level, constitutive expression
- C-terminal V5 epitope tag for efficient detection of recombinant proteins with an Anti-V5 antibody
- C-terminal polyhistidine (6xHis) sequence for purification using nickel-chelating resin and detection with Anti-His (C-term)antibody
- Topoisomerase activation of pcDNA3.1/V5-His-TOPO vector allows PCR products to be ligated in 5 minutes on bench top and results in 90% recombinants
- Vector features strong CMV promoter for high-level, constitutive expression
- C-terminal V5 epitope tag for efficient detection of recombinant proteins with Anti-V5 antibody
Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Dry Ice
Specifications
Constitutive | |
CMV | |
Geneticin™ (G-418) | |
TOPO-TA | |
TOPO™, pcDNA™ |
Transfection | |
TOPO TA Expression Kit | |
His Tag (6x), V5 Epitope Tag | |
TOPO-TA Vectors, pcDNA | |
Constitutive Expression |
For Research Use Only. Not for use in diagnostic procedures.