Learn More
Invitrogen™ pBAD TOPO™ TA Expression Kit
Product Code. 10115092
Change view
Click to view available options
Protein Tag:
His Tag (6x), V5 Epitope Tag
V5 Epitope Tag
Quantity:
20 Reactions
40 Reactions
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Protein Tag Quantity unitSize
10115092 His Tag (6x), V5 Epitope Tag 20 Reactions Each
10414592 V5 Epitope Tag 40 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 10115092 Supplier Invitrogen™ Supplier No. K430001
For Research Use Only. All usage must comply with product instructions.
Includes
Each pBAD TOPO TA Expression Kit contains two boxes and the LMG194 E. coli stab. The pBAD TOPO TA box contains all of the reagents necessary for PCR (except Taq polymerase), including 200ng of topoisomerase I-activated pBAD-TOPO vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control template and primers, 20% L-arabinose, pBAD primers for sequencing or PCR screening, and an expression control plasmid. The other box contains all of the reagents necessary for transformation, including single-use 50μL aliquots of TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid.
Specifically designed for one-step cloning and regulated prokaryotic expression of Taq-amplified PCR products
  • Linearized, topoisomerase I-activated vector for 5-minute cloning of Taqamplified PCR products
  • araBAD promoter for tightly regulated expression in E. coli V5 epitope tag for detection with Anti-V5 Antibody
  • C-terminal polyhistidine (6xHis) tag for purification using nickel-chelating resin and detection with Anti-His(C-term) Antibody
  • Enterokinase cleavage site for removal of N-terminal leader peptide
Bacterial Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Constitutive or Inducible System Inducible
Inducing Agent Arabinose
Promoter araBAD
Product Type TA Expression Kit
Selection Agent (Eukaryotic) None
Content And Storage Each pBAD TOPO™ TA Expression Kit contains two boxes and the LMG194 E. coli stab. The pBAD TOPO™ TA box contains all of the reagents necessary for PCR (except Taq polymerase), including 200 ng of topoisomerase I-activated pBAD-TOPO™ vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control template and primers, 20% L-arabinose, pBAD primers for sequencing or PCR screening, and an expression control plasmid. Store at -20°C. The One Shot™ box contains all of the reagents necessary for transformation, including single-use 50 μl aliquots of One Shot™ TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Store One Shot™ cells at -80°C. Store the LMG194 E. coli stab at 2 - 8°C. All reagents are guaranteed stable for 6 months when properly stored.
Antibiotic Resistance Bacterial Ampicillin (AmpR)
Bacterial or Yeast Strain LMG194, TOP10
Cleavage EK (Enterokinase) Recognition Site
Expression System E. coli
Expression Mechanism Cell-Based Expression
Protein Tag His Tag (6x), V5 Epitope Tag
Cloning Method TOPO-TA
Quantity 20 Reactions
Vector pBAD
Product Line TOPO