missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ TOP10/P3 Chemically Competent E. coli

Product Code. 10719384
Change view
Click to view available options
Quantity:
21 x 50 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10719384 21 x 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 10719384 Supplier Invitrogen™ Supplier No. C505003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

One Shot TOP10/P3 Chemically Competent E. coli cells are designed for vector transformation encoding the synthetic supF gene (tyrosine tRNA suppressor) and allow high-quality pCDM8, pcDNA1.1 plasmid, or any other supF-containing vector preparation.

One Shot TOP10/P3 Chemically Competent E. coli cells are designed for vector transformation encoding the synthetic supF gene (tyrosine tRNA suppressor) and allow high-quality pCDM8, pcDNA1.1 plasmid, or any other supF-containing vector preparation.

One Shot TOP10/P3 strain features
• P3 plasmid for selection of supF-containing plasmids
• Mutated endA1 gene for increased quality of plasmid DNA preparations
recA1 for reduced occurrence of non-specific recombination in cloned DNA
lacZΔM15 for blue/white color screening of recombinant clones

P3 Plasmid
The P3 plasmid is a low-copy-number, 60 kb plasmid that carries kanamycin, tetracycline and ampicillin resistance markers. The kanamycin gene is fully active and is used to select for cells carrying P3. Tetracycline and ampicillin genes carry amber mutations which render the genes inactive during normal growth and replication of the bacteria. Upon transformation of a vector carrying the suppressor F gene, the amber mutations in the tetracycline and ampicillin genes on the P3 plasmid are suppressed and the E. coli are resistant to these antibiotics.

Genotype
F- mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK rpsL endA1 nupG P3: KanR AmpR (am) TetR (am) 

Find the strain and format that you need
• TOP10 is offered in chemically competent and electrocompetent cell formats.
• Choose MultiShot competent cells for high-throughput cloning applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level Medium Efficiency (1 x 108 to 1 x 109 cfu/μg)
Content And Storage • One Shot TOP10/P3 Chemically Competent E. coli (21 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 Plasmid (50 μL)
Store pUC19 plasmid at –20°C.

S.O.C. Medium (6 mL)
Store S.O.C. Medium at 4°C or at room temperature
Antibiotic Resistance Bacterial Yes (Streptomycin, Kanamycin, Ampicillin, Tetracycline)
Cloning Unstable DNA No
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Format Tube
Product Line One Shot
Quantity 21 x 50 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.