missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NativePure™ Lentiviral Gateway™ Vector Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
Includes
pLenti6/capTEV-NT-DEST1 (N-terminal vector), pLenti6/capTEV-CT-DEST (C-terminal vector), and pLenti6/capTEV-CT-GW/ARP2 (C-terminal) control vector.
- Allow N- or C- terminal fusion of recombinant protein of interest to capTEV± affinity purification tag
- Gateway vectors designed to express protein of interest in multiple expression systems
- Lentiviral vectors with capTEVTag that allow in vivo biotinylation of protein of interest
- Streamlined purification protocols with Streptavidin Agarose
Protein and Peptide Purification,Protein Complex Isolation and Analysis,Protein Sample Preparation and Protein Purification,Protein-Protein Interactions,Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Constitutive or Inducible System | Constitutive |
| Delivery Type | Lentiviral |
| Promoter | CMV |
| Selection Agent (Eukaryotic) | Blasticidin |
| Content And Storage | Store the 293FT Cell Line in liquid nitrogen and the One Shot Stbl3 Chemically Competent E. coli at -80°C. Store Streptavidin Agarose, 10% NP-40, NativePure± 5X Lysis/Binding Buffer, NativePure± 10X TEV Cleavage Buffer, Lipofectamine 2000, NativePure± Columns, and NativePure± Concentrators at +4°C. Store the NativePure Lentiviral Gateway Vector Kit, AcTEV± Protease, 100mM DTT, Gateway LR Clonase Enzyme Mix, ViraPower packaging Mix, and Blasticidin at -20°C. All components are guaranteed stable for 6 months when properly stored. |
| Protein Tag | BioEase Tag, His Tag (6x), V5 Epitope Tag |
| Quantity | 1 Kit |
| Vector | pLenti |
| For Use With (Application) | Protein-Protein Interaction Studies, Viral Expression |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction