missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mifepristone
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Includes
100μg mifepristone is provided lyophilized.
- Subphysiological levels of steroid compound are used to induce expression from very low basal to high induced levels
- Formulated for use in GeneSwitch™ System
- Lypholized
Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression
Specifications
Specifications
| Format | Lyophilized |
| Product Type | Mifepristone |
| Content And Storage | 100 μg mifepristone is provided lyophilized. Store at -20°C. Mifepristone is guaranteed stable for 6 months when properly stored. |
| Quantity | 100 μg |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction