missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mammalian Lumio™ Gateway™ Vectors, with Lumio™ Green In-Cell Detection Kit

Product Code. 10739803
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10739803

Brand: Invitrogen™ 12589016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

All destination vectors are provided lyophilized and supercoiled.

Requires

Additional materials required, available separately: Gateway™ entry clone, Gateway™ LR Clonase™ enzyme mix, and reaction buffer.

Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or Ultimate™ RF Clone

Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Cloning, Constitutive Expression, Enzyme and Protein Activity Assays, Gateway Cloning, Lumio Detection, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Reporter Assays, Tetracysteine (Lumio/FlAsH/ReAsH) Assays

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Transfection
Promoter CMV
Product Type Vector with In-cell Detection Kit
Selection Agent (Eukaryotic) Blasticidin
Content And Storage All destination vectors are provided lyophilized and supercoiled.
Detection Location In-Cell Detection
Protein Tag Lumio, V5 Epitope Tag
Cloning Method Gateway
Quantity 1 kit
Vector pcDNA
Product Line Gateway, Lumio
For Use With (Application) Reporter Assays
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.