missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagnaRack™ Magnetic Separation Rack

Product Code. 10625463
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10625463 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10625463 Supplier Invitrogen™ Supplier No. CS15000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MagnaRack consists of magnetic base station and removable tube rack.

  • MagnaRack consists of magnetic base station and removable tube rack
  • Tube rack fits onto magnetic base station in two different positions, associating row of 12 neodinium magnets with single row of 12 tubes for easy 'on the magnet' and 'off the magnet' sample processing

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
For Use With (Equipment) MagnaRack™ Magnetic Separator
Shipping Condition Room Temperature
Product Line MagnaRack
Quantity 1 Each
Type Magnetic Separation Rack

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.