missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 24-Well Magnetic Separator

Product Code. 10309022
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10309022

Brand: Invitrogen™ CS15024

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

One Magnetic Separator

Contains six neodymium magnets that aligned with wells of a 24-well plate for efficient processing of DNA bound to magnetic beads.

  • Contains six neodymium magnets that aligned with wells of a 24-well plate for efficient processing of DNA bound to magnetic beads
  • GeneCatcher technology is novel magnetic bead-based method that reliably and efficiently purifying DNA without any centrifugation steps
  • GeneCatcher gDNA Blood Kits facilitate efficient and reliable extraction of genomic DNA (gDNA) from samples of human blood, even when the samples have been archived, poorly stored, or collected in the presence of anticoagulants such as EDTA, heparin, or citrate

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one Magnetic Separator, which may be stored at room temperature.
For Use With (Equipment) MagnaRack™ Magnetic Separator, Magna-Sep™ Magnetic Particle Separator
Shipping Condition Room Temperature
Product Line MagnaRack
Quantity 1 Ea.
Type Magnetic Separator

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.