missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ M13KO7 Helper Phage

Product Code. 10433022
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10433022 5 x 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10433022 Supplier Invitrogen™ Supplier No. 18311019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Suitable for use with M13/phagemid hosts for the production of ssDNA from vectors containing an f1or f1-like origin of replication

Prepared by modification of procedure of Vieira and Messing, this high-titer helper phage eliminates need to make and maintain a separate stock of helper phage. It contains Kmr gene and p15A origin of replication that allows it to replicate like plasmid inside cells. Detailed protocols for production of ssDNA are provided on product profile.

  • Convenient preparation of ssDNA from M13 or phagemid vectors
  • >1 x 1011 pfu/mL

Cloning, Mutagenesis

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in ultra-cold freezer (-68 to -85°C).
Shipping Condition Dry Ice
Quantity 5 x 1 mL
Product Type M13KO7 Helper Phage

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.