missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lumio™ Green In-Cell Detection Kit

Product Code. 10388152
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10388152 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10388152 Supplier Invitrogen™ Supplier No. 12589057

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

6μg pcDNA 6.2/nLumio;-DEST, 6μg pcDNA 6.2/cLumio-DEST, 10μg pcDNA6.2/nLumio;-GW/p64 control vector, and the indicated Lumio In-Cell Detection Kit. The Lumio In-Cell Detection Kits include 40μL of Lumio Green or Red (Dual contains both Green and Red) fluorescent reagent as a 2mM stock solution in DMSO.

Designed for simple fluorescent detection of recombinant protein

  • Tag is small (six amino acid) sequence that binds fluorescent substrate, allowing to visually detect protein localization in mammalian cells
  • Offers accurate in vitro and in vivo protein detection
  • CMV promoter for high-level constitutive expression
  • tR sites for efficient recombination with Gateway entry clones
  • Blasticidin resistance gene for fast, efficient selection
  • Use for reliable and consistent protein expression, detection, and localization in mammalian cells

Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Enzyme and Protein Activity Assays, Lumio Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays, Tetracysteine (Lumio/FlAsH/ReAsH) Assays

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Electron Microscope
Product Type In-Cell Detection Kit
Content And Storage The Lumio In-Cell Detection kits include 40 μL of Lumio Green or Red fluorescent reagent as a 2 mM stock solution in DMSO. Store at -20°C. Guaranteed stable for 6 months when properly stored.
Detection Location In-Cell Detection
Protein Tag Lumio
Label or Dye Lumio™ Green
Quantity 1 kit
Product Line Lumio
Color Green

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.