Learn More
Invitrogen™ iPrep™ PureLink™ gDNA Blood Kit
Description
Includes
The iPrep™ PureLink™ gDNA Blood Kit contains enough material to perform 52 purifications. Contains: 1 iPrep™ PureLink™ gDNA Blood Cartridge Kit, 2 x 52 iPrep™ Sample and Elution Tubes, 1 bag iPrep™ Tips and Tip Holders (52 tips and holders)
Genomic DNA is extracted from blood samples using the Dynabeads™ MyOne™ SILANE and iPrep™ Purification Instrument within 30 minutes without the use of centrifugation. The purified genomic DNA is suitable for use in downstream applications, including PCR.
Using the iPrep™ PureLink™ gDNA Blood Kit
With A260/A280 ratios of 1.8, the resulting DNA is highly pure, ideal for use in sensitive multiplex assays or for long-term storage. This kit is optimized to isolate highly-pure DNA from 150μL to 350μL of fresh or frozen blood in less than 30 minutes. It is highly tolerant of anticoagulants such as citrate and EDTA, and yields high molecular weight DNA. The yield is highly reproducible, and there is no cross-contamination between samples. Furthermore, the kit can also be used with cultured cells. The table summarizes the performance of the iPrep™ PureLink™ gDNA Blood Kit. The iPrep™ PureLink™ gDNA Blood Kit comes with all reagents (supplied in prefilled cartridges), tubes, and tips needed to isolate DNA from blood samples. All you need to do is add the cartridges, tips, and your sample tubes to the instrument, select your preferred elution volume, and come back 30 minutes later to collect your pure gDNA in individual screwcap elution tubes.
- Obtain high yields of gDNA from up to 350μL blood
- Isolate highly pure gDNA with high A260/A280 and A260/A230 ratios
- Experience superior performance in sensitive downstream applications
- Purify high-quality gDNA from fresh or frozen whole blood
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | The iPrep™ PureLink™ gDNA Blood Kit contains enough material to perform 52 purifications Contains:
|
| Isolation Technology | Magnetic Bead |
| Sample Type | Blood |
| Final Product Type | Genomic DNA |
| For Use With (Application) | PCR, Southern Blotting, Sequencing |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Includes | 1 iPrep™ PureLink™ gDNA Blood Cartridge Kit; 2 × 52 iPrep™ Sample and Elution Tubes; 1 bag iPrep™ Tips and Tip Holders (52 tips and holders) |
| No. of Reactions | 52 |
| Prep Scale | 100 μl - 1 ml Blood Samples ( Medium-Scale) |
| Product Line | PureLink, iPrep |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.