missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tfi DNA Polymerase
Specifications
Specifications
| Content And Storage | Tfi DNA Polymerase is supplied with a vial of magnesium-free 5X Tfi PCR Buffer [250mM TrisHCl (pH 8.4), 75mM (NH4)2SO4, 25mM KCl, proprietary stabilizers, 1mM DTT], and a vial of 50mM MgCl2. Store at -20°C. Guaranteed stable for 6 months when properly stored. |
| GC-Rich PCR Performance | Low |
| Polymerase | Tfi DNA Polymerase |
| Exonuclease Activity | 5' - 3' |
| Quantity | 500 Reactions |
| Fidelity (vs. Taq) | 1 X |
| No. of Reactions | 500 |
| Overhang | 3'-A |
| Size (Final Product) | 4 kb or less |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?